3-Amino-7-fluoroquinoline dihydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 7-Fluoroquinolin-3-amine dihydrochloride

PC49035-1G
APOSPC49035-1GEA 0 EUR
APOSPC49035-1G
3-Amino-7-fluoroquinoline dihydrochloride
3-Amino-7-fluoroquinoline dihydrochloride
Formel: C₉H₇FN₂·2ClH
Molecular Weight: 235,086 g/mol
Schmelzpunkt: 185…186 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD18375333
CAS-Nummer: 1266226-28-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR