3-(Aminomethyl)-5-(trifluoromethyl)pyridine dihydrochloride 95%

Lieferant: Apollo Scientific
Achtung

PC49039-1G PC49039-500MG
APOSPC49039-1GEA 0 EUR
APOSPC49039-1G APOSPC49039-500MG
3-(Aminomethyl)-5-(trifluoromethyl)pyridine dihydrochloride 95%
3-(Aminomethyl)-5-(trifluoromethyl)pyridine dihydrochloride
Formel: C₇H₇F₃N₂·2ClH
Schmelzpunkt: 210 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD16988625
CAS-Nummer: 1020747-92-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR