5-Chloro-3-(3-fluorobenzyl)-1,2,4-thiadiazole

Lieferant: Apollo Scientific

Synonyme: 1-[(5-Chloro-1,2,4-thiadiazol-3-yl)methyl]-3-fluorobenzene, 5-Chloro-3-[(3-fluorophenyl)methyl]-1,2,4-thiadiazole

PC49046-1G
APOSPC49046-1GEA 517 EUR
APOSPC49046-1G
5-Chloro-3-(3-fluorobenzyl)-1,2,4-thiadiazole
5-Chloro-3-(3-fluorobenzyl)-1,2,4-thiadiazole
Formel: C₉H₆ClFN₂S
Molecular Weight: 228,674 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14707331
CAS-Nummer: 1029718-68-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR