Methyl-(4-fluorphenyl)acetat 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 2-(4-fluorophenyl)acetate

PC49048-100G
APOSPC49048-100GEA 112 EUR
APOSPC49048-100G
Methyl-(4-fluorphenyl)acetat 98%
Methyl-(4-fluorphenyl)acetat
Formel: C₉H₉FO₂
Molecular Weight: 168,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00800608
CAS-Nummer: 34837-84-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR