Phenyl trifluoromethyl sulphone 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Phenyl Trifluoromethyl sulphone

PC49074-5G
APOSPC49074-5GEA 119 EUR
APOSPC49074-5G
Phenyl trifluoromethyl sulphone 98%
Phenyl trifluoromethyl sulphone
Formel: C₇H₅F₃O₂S
Dichte: 1,423 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00159083
CAS-Nummer: 426-58-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR