2'-Fluoro-3'-methoxyacetophenone 97%

Lieferant: Apollo Scientific
Achtung

PC49075-1G PC49075-250MG PC49075-5G
APOSPC49075-1GEA 270 EUR
APOSPC49075-1G APOSPC49075-250MG APOSPC49075-5G
2'-Fluoro-3'-methoxyacetophenone 97%
2'-Fluoro-3'-methoxyacetophenone
Formel: C₉H₉FO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD13194330
CAS-Nummer: 208777-19-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR