2-Fluoro-4-nitrobenzyl bromide 97%

Lieferant: Apollo Scientific
Gefahr

PC49076-1G PC49076-250MG PC49076-5G
APOSPC49076-1GEA 0 EUR
APOSPC49076-1G APOSPC49076-250MG APOSPC49076-5G
2-Fluoro-4-nitrobenzyl bromide 97%
2-Fluoro-4-nitrobenzyl bromide
Formel: C₇H₅BrFNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD10000853
CAS-Nummer: 127349-56-8
UN: 3261
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR