4-Bromo-2-methoxybenzotrifluoride

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-2-(trifluoromethyl)anisole, 5-Bromo-2-(trifluoromethyl)phenyl methyl ether, 4-Bromo-2-methoxy-1-(trifluoromethyl)benzene

PC49083-1G PC49083-25G PC49083-5G
APOSPC49083-1GEA 31.9 EUR
APOSPC49083-1G APOSPC49083-25G APOSPC49083-5G
4-Bromo-2-methoxybenzotrifluoride
4-Bromo-2-methoxybenzotrifluoride
Formel: C₈H₆BrF₃O
Molecular Weight: 255.032 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13194486
CAS-Nummer: 944805-63-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR