3-Bromo-6-fluoropyridine-2-carboxylic acid

Lieferant: Apollo Scientific

PC49090-250MG
APOSPC49090-250MGEA 84.4 EUR
APOSPC49090-250MG
3-Bromo-6-fluoropyridine-2-carboxylic acid
3-Bromo-6-fluoropyridine-2-carboxylic acid
Formel: C₆H₃BrFNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD13185802
CAS-Nummer: 1211589-43-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR