(Trifluoromethoxy)acetic acid

Lieferant: Apollo Scientific
Achtung

PC49092-1G PC49092-500MG
APOSPC49092-1GEA 415 EUR
APOSPC49092-1G APOSPC49092-500MG
(Trifluoromethoxy)acetic acid
(Trifluoromethoxy)acetic acid
Formel: C₃H₃F₃O₃
Siedepunkt: 84 °C (33 mm Hg)
Dichte: 1,5014 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD11858185
CAS-Nummer: 69105-00-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR