1-Chloro-7-fluoroisoquinoline

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Chloro-7-fluoro-2-azanaphthalene

PC49097-1G PC49097-500MG
APOSPC49097-1GEA 531 EUR
APOSPC49097-1G APOSPC49097-500MG
1-Chloro-7-fluoroisoquinoline
1-Chloro-7-fluoroisoquinoline
Formel: C₉H₅ClFN
Molecular Weight: 181.594 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16883053
CAS-Nummer: 630422-89-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR