1-Chloro-7-fluoroisoquinoline
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Chloro-7-fluoro-2-azanaphthalene
PC49097-1G
PC49097-500MG
APOSPC49097-1GEA
531
EUR
APOSPC49097-1G
APOSPC49097-500MG
1-Chloro-7-fluoroisoquinoline
1-Chloro-7-fluoroisoquinoline
Formel:
C₉H₅ClFN Molecular Weight: 181.594 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16883053 CAS-Nummer: 630422-89-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...