1,3-Diisopropylimidazoliumtetrafluoroborat
Lieferant: Apollo Scientific
|
Achtung
|
PC49102-5G
APOSPC49102-5GEA
201
EUR
APOSPC49102-5G
1,3-Diisopropylimidazoliumtetrafluoroborat
1,3-Diisopropylimidazoliumtetrafluoroborat
Formel:
C₉H₁₇BF₄N₂ Schmelzpunkt: 62…79 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08276804 CAS-Nummer: 286014-34-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...