2,4,6-Trifluorophenylsulphur pentafluoride

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Pentafluorothio)-1,3,5-trifluorobenzene, 2-(Pentafluorosulphanyl)-1,3,5-trifluorobenzene

PC49106-1G
APOSPC49106-1GEA 437 EUR
APOSPC49106-1G
2,4,6-Trifluorophenylsulphur pentafluoride
2,4,6-Trifluorophenylsulphur pentafluoride
Formel: C₆H₂F₈S
Molecular Weight: 258,132 g/mol
Siedepunkt: 140 °C (637 mm Hg)
Lagertemperatur: Raumtemperatur
MDL: MFCD18071066
CAS-Nummer: 1062610-12-1
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR