1,4-Bis(pentafluorothio)benzene
Lieferant: Apollo Scientific
PC49108-1G
APOSPC49108-1GEA
1060
EUR
APOSPC49108-1G
1,4-Bis(pentafluorothio)benzene
1,4-Bis(pentafluorothio)benzene
Formel:
C₆H₄F₁₀S₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD18071068 CAS-Nummer: 1219501-58-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...