1,4-Bis(pentafluorothio)perfluorobenzene
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
1,4-Bis(pentafluorosulphanyl)-2,3,5,6-tetrafluorobenzene, 1,4-Bis(pentafluorothio)-2,3,5,6-tetrafluorobenzene
PC49109-1G
APOSPC49109-1GEA
538
EUR
APOSPC49109-1G
1,4-Bis(pentafluorothio)perfluorobenzene
1,4-Bis(pentafluorothio)perfluorobenzene
Formel:
C₆F₁₄S₂ Schmelzpunkt: 129…130 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD18071069 CAS-Nummer: 1219501-60-6 UN: 1325 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...