5-Fluoroquinoline

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Fluoro-1-azanaphthalene

PC49118-1G
APOSPC49118-1GEA 524 EUR
APOSPC49118-1G
5-Fluoroquinoline
5-Fluoroquinoline
Formel: C₉H₆FN
Molecular Weight: 147,149 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01685511
CAS-Nummer: 394-69-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR