(2,4-Difluorophenyl)methanesulphonyl chloride

Lieferant: Apollo Scientific
Gefahr

PC49122-1G PC49122-500MG
APOSPC49122-1GEA 0 EUR
APOSPC49122-1G APOSPC49122-500MG
(2,4-Difluorophenyl)methanesulphonyl chloride
(2,4-Difluorophenyl)methanesulphonyl chloride
Formel: C₇H₅ClF₂O₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD06798046
CAS-Nummer: 179524-68-6
UN: 3265
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR