2-Ethynyl-5-(trifluoromethyl)pyridine

Lieferant: Apollo Scientific
Achtung

Synonyme: [5-(Trifluoromethyl)pyridin-2-yl]acetylene, 6-Ethynyl-α,α,α-trifluoro-3-picoline

PC49133-25MG
APOSPC49133-25MGEA 21.9 EUR
APOSPC49133-25MG
2-Ethynyl-5-(trifluoromethyl)pyridine
2-Ethynyl-5-(trifluoromethyl)pyridine
Formel: C₈H₄F₃N
Molecular Weight: 171,119 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13189717
CAS-Nummer: 379670-42-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR