4-[(2-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]amino}ethyl)amino]-4-oxo-2-butenoic acid

Lieferant: Apollo Scientific

PC4914-5G PC4914-500MG PC4914-1G
APOSPC4914-5GEA 1510 EUR
APOSPC4914-5G APOSPC4914-500MG APOSPC4914-1G
4-[(2-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]amino}ethyl)amino]-4-oxo-2-butenoic acid
4-[(2-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]amino}ethyl)amino]-4-oxo-2-butenoic acid
Formel: C₁₂H₁₁ClF₃N₃O₃
Schmelzpunkt: 166…167 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00202686
CAS-Nummer: 339096-60-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR