Methyltrifluoracetat 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: Trifluoroacetic methyl ester

PC49140-1KG
APOSPC49140-1KGEA 0 EUR
APOSPC49140-1KG
Methyltrifluoracetat 97%
Methyltrifluoracetat

Useful intermediate for trifluoroacetylation of amines. Can also be used for the N-acylation of amino acids (with1,1,3,3-tetramethylguanidine): Synthesis, 399 (1976)

Formel: C₃H₃F₃O₂
Molecular Weight: 128,05 g/mol
Siedepunkt: 43…44 °C
Schmelzpunkt: –78 °C
Dichte: 1,283 g/cm³ (25 °C)
Flammpunkt: –18 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00000417
CAS-Nummer: 431-47-0
EINECS: 207-074-5
UN: 2924
ADR: 3+8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR