5-Ethynyl-2-fluoropyridine 97%

Lieferant: Apollo Scientific

PC49141-1G
APOSPC49141-1GEA 0 EUR
APOSPC49141-1G
5-Ethynyl-2-fluoropyridine 97%
5-Ethynyl-2-fluoropyridine
Formel: C₇H₄FN
Lagertemperatur: Kühlschrank
MDL: MFCD13175289
CAS-Nummer: 853909-08-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR