3-(Difluoromethyl)aniline 95%

Lieferant: Apollo Scientific
Achtung

PC49148-1G PC49148-250MG PC49148-5G
APOSPC49148-1GEA 0 EUR
APOSPC49148-1G APOSPC49148-250MG APOSPC49148-5G
3-(Difluoromethyl)aniline 95%
3-(Difluoromethyl)aniline
Formel: C₇H₇F₂N
Lagertemperatur: Raumtemperatur
MDL: MFCD18384830
CAS-Nummer: 368-99-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR