2-{2-[3-Chloro-5-(trifluoromethyl)pyridin-2-ylamino]ethyl}amino-5-(trifluoromethyl)aniline hydrochloride

Lieferant: Apollo Scientific

PC4915-500MG
APOSPC4915-500MGEA 783 EUR
APOSPC4915-500MG
2-{2-[3-Chloro-5-(trifluoromethyl)pyridin-2-ylamino]ethyl}amino-5-(trifluoromethyl)aniline hydrochloride
2-{2-[3-Chloro-5-(trifluoromethyl)pyridin-2-ylamino]ethyl}amino-5-(trifluoromethyl)aniline hydrochloride
Formel: C₁₅H₁₃ClF₆N₄·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD00793061
CAS-Nummer: 317822-50-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR