4-Methoxy-2-(trifluoromethoxy)benzaldehyde

Lieferant: Apollo Scientific
Achtung

PC49158-10G PC49158-5G
APOSPC49158-10GEA 0 EUR
APOSPC49158-10G APOSPC49158-5G
4-Methoxy-2-(trifluoromethoxy)benzaldehyde
4-Methoxy-2-(trifluoromethoxy)benzaldehyde
Formel: C₉H₇F₃O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD06660317
CAS-Nummer: 886503-52-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR