3-Brom-5-fluor-2-methoxypyridin 98%

Lieferant: Apollo Scientific
Achtung

PC49160-25G
APOSPC49160-25GEA 81.2 EUR
APOSPC49160-25G
3-Brom-5-fluor-2-methoxypyridin 98%
3-Bromo-5-fluoro-2-methoxypyridine
Formel: C₆H₅BrFNO
Lagertemperatur: Kühlschrank
MDL: MFCD04972397
CAS-Nummer: 884494-81-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR