2,5-Difluoro-4-hydrazinylbenzonitrile 95+%

Lieferant: Apollo Scientific

PC49169-250MG
APOSPC49169-250MGEA 102 EUR
APOSPC49169-250MG
2,5-Difluoro-4-hydrazinylbenzonitrile 95+%
2,5-Difluoro-4-hydrazinylbenzonitrile
Formel: C₇H₅F₂N₃
Molecular Weight: 169,13 g/mol
Schmelzpunkt: 159…161 °C
Lagertemperatur: Kühlschrank
MDL: MFCD04972812
CAS-Nummer: 129946-63-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR