4-Brom-2-methylbenzotrifluorid
Lieferant: Apollo Scientific
Synonyme:
4-Bromo-2-methyl-1-(trifluoromethyl)benzene
PC49171-1G
APOSPC49171-1GEA
26.1
EUR
APOSPC49171-1G
4-Brom-2-methylbenzotrifluorid
4-Brom-2-methylbenzotrifluorid
Formel:
C₈H₆BrF₃ Molecular Weight: 239,0325 g/mol |
MDL:
MFCD11100526 CAS-Nummer: 936092-88-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...