4-Bromo-1-(difluoromethyl)-2-fluorobenzene 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromo-2-fluorobenzal fluoride

PC49172-25G
APOSPC49172-25GEA 561 EUR
APOSPC49172-25G
4-Bromo-1-(difluoromethyl)-2-fluorobenzene 97%
4-Bromo-1-(difluoromethyl)-2-fluorobenzene
Formel: C₇H₄BrF₃
Molecular Weight: 225,01 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06657963
CAS-Nummer: 749932-17-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR