4-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]amino}-3-hydroxybutanenitrile 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]amino}-3-cyanopropan-2-ol, 3-Chloro-2-[(3-cyano-2-hydroxyprop-1-yl)amino]-5-(trifluoromethyl)pyridine
PC4919-500MG
APOSPC4919-500MGEA
749
EUR
APOSPC4919-500MG
4-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]amino}-3-hydroxybutanenitrile 97%
4-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]amino}-3-hydroxybutanenitrile
Formel:
C₁₀H₉ClF₃N₃O Molecular Weight: 279,646 g/mol Schmelzpunkt: 107…109 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01315213 CAS-Nummer: 338966-48-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...