2-Amino-5-fluoro-1,3-thiazole hydrochloride 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Fluoro-1,3-thiazol-2-amine hydrochloride

PC49194-1G
APOSPC49194-1GEA 158 EUR
APOSPC49194-1G
2-Amino-5-fluoro-1,3-thiazole hydrochloride 97%
2-Amino-5-fluoro-1,3-thiazole hydrochloride
Formel: C₃H₃FN₂S·ClH
Molecular Weight: 154,594 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09056854
CAS-Nummer: 745053-64-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR