3-(Difluoromethyl)benzonitrile

Lieferant: Apollo Scientific
Achtung

PC49196-1G PC49196-5G PC49196-25G
APOSPC49196-1GEA 26.1 EUR
APOSPC49196-1G APOSPC49196-5G APOSPC49196-25G
3-(Difluoromethyl)benzonitrile
3-(Difluoromethyl)benzonitrile
Formel: C₈H₅F₂N
Molecular Weight: 153.13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19053256
CAS-Nummer: 55805-13-5
UN: 3276
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR