3-(Difluoromethyl)benzaldehyde 97%

Lieferant: Apollo Scientific
Achtung

PC49198-1G PC49198-5G
APOSPC49198-1GEA 0 EUR
APOSPC49198-1G APOSPC49198-5G
3-(Difluoromethyl)benzaldehyde 97%
3-(Difluoromethyl)benzaldehyde
Formel: C₈H₆F₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD18791599
CAS-Nummer: 945004-44-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR