3-(Difluoromethyl)benzyl bromide 97%

Lieferant: Apollo Scientific
Gefahr

PC49200-5G PC49200-1G PC49200-25G
APOSPC49200-5GEA 0 EUR
APOSPC49200-5G APOSPC49200-1G APOSPC49200-25G
3-(Difluoromethyl)benzyl bromide 97%
3-(Difluoromethyl)benzyl bromide
Formel: C₈H₇BrF₂
Lagertemperatur: Kühlschrank
MDL: MFCD18791600
CAS-Nummer: 1263178-51-3
UN: 3265
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR