4-Hydroxy-6-(trifluoromethyl)pyrimidine 97%

Lieferant: Apollo Scientific
Achtung

PC49210-5G
APOSPC49210-5GEA 21.9 EUR
APOSPC49210-5G
4-Hydroxy-6-(trifluoromethyl)pyrimidine 97%
4-Hydroxy-6-(trifluoromethyl)pyrimidine
Formel: C₅H₃F₃N₂O
Molecular Weight: 164,09 g/mol
Schmelzpunkt: 169…171 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00192530
CAS-Nummer: 1546-78-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR