2,3-Dichloro-5-fluoropyridine 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC49213-5G
APOSPC49213-5GEA
102
EUR
APOSPC49213-5G
2,3-Dichloro-5-fluoropyridine 98%
2,3-Dichloro-5-fluoropyridine
Formel:
C₅H₂Cl₂FN Lagertemperatur: Raumtemperatur |
MDL:
MFCD08669666 CAS-Nummer: 185985-40-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...