[5-({[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]amino}methyl)-4,5-dihydro-3-isoxazolyl]methanol 97%

Lieferant: Apollo Scientific

Synonyme: 5-[3-Chloro-5-(trifluoromethyl)pyridin-2-ylamino]methyl-4,5-dihydro-3-(hydroxymethyl)isoxazole

PC4922-500MG
APOSPC4922-500MGEA 783 EUR
APOSPC4922-500MG
[5-({[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]amino}methyl)-4,5-dihydro-3-isoxazolyl]methanol 97%
[5-({[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]amino}methyl)-4,5-dihydro-3-isoxazolyl]methanol
Formel: C₁₁H₁₁ClF₃N₃O₂
Molecular Weight: 309,68 g/mol
Schmelzpunkt: 112…114 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00202658
CAS-Nummer: 338410-31-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR