5-Amino-3-fluoropyridine-2-carbonitrile 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Amino-3-fluoropicolinonitrile, 5-Amino-2-cyano-3-fluoropyridine

PC49229-250MG
APOSPC49229-250MGEA 241 EUR
APOSPC49229-250MG
5-Amino-3-fluoropyridine-2-carbonitrile 95%
5-Amino-3-fluoropyridine-2-carbonitrile
Formel: C₆H₄FN₃
Molecular Weight: 137,114 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12911860
CAS-Nummer: 573763-07-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR