4-(Trifluoromethoxy)phenoxyacetyl chloride

Lieferant: Apollo Scientific
Gefahr

PC4923-1G PC4923-5G
APOSPC4923-1GEA 0 EUR
APOSPC4923-1G APOSPC4923-5G
4-(Trifluoromethoxy)phenoxyacetyl chloride
4-(Trifluoromethoxy)phenoxyacetyl chloride
Formel: C₉H₆ClF₃O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD02091505
CAS-Nummer: 72220-51-0
UN: 1760
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR