3-Amino-5-fluoropyridine-2-carbonitrile
Lieferant: Apollo Scientific
|
Achtung
|
PC49230-250MG
APOSPC49230-250MGEA
0
EUR
APOSPC49230-250MG
3-Amino-5-fluoropyridine-2-carbonitrile
3-Amino-5-fluoropyridine-2-carbonitrile
Formel:
C₆H₄FN₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD20259119 CAS-Nummer: 1374115-60-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...