5-Bromo-2-hydrazino-3-(trifluoromethyl)pyridine

Lieferant: Apollo Scientific
Achtung

Synonyme: [5-Bromo-3-(trifluoromethyl)pyridin-2-yl]hydrazine, 1-(5-bromo-3-(trifluoromethyl)pyridin-2-yl)hydrazine, 5-Bromo-2-hydrazino-α,α,α-trifluoro-3-picoline

PC49235-1G PC49235-250MG
APOSPC49235-1GEA 255 EUR
APOSPC49235-1G APOSPC49235-250MG
5-Bromo-2-hydrazino-3-(trifluoromethyl)pyridine
5-Bromo-2-hydrazino-3-(trifluoromethyl)pyridine
Formel: C₆H₅BrF₃N₃
Lagertemperatur: Kühlschrank
MDL: MFCD22375529
CAS-Nummer: 1374652-48-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR