Methyl 4-fluoro-2-(methoxymethyl)-5-nitrobenzoate

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Fluoro-5-(methoxycarbonyl)-4-(methoxymethyl)nitrobenzene

PC49238-1G
APOSPC49238-1GEA 65.4 EUR
APOSPC49238-1G
Methyl 4-fluoro-2-(methoxymethyl)-5-nitrobenzoate
Methyl 4-fluoro-2-(methoxymethyl)-5-nitrobenzoate
Formel: C₁₀H₁₀FNO₅
Molecular Weight: 243.189 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22378816
CAS-Nummer: 1243164-13-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR