5-[1-Methyl-5-(trifluoromethyl)-1H-pyrazol-3-yl]thiophene-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(5-Carboxythien-2-yl)-1-methyl-5-(trifluoromethyl)-1H-pyrazole

PC4924-100MG PC4924-1G
APOSPC4924-100MGEA 72.5 EUR
APOSPC4924-100MG APOSPC4924-1G
5-[1-Methyl-5-(trifluoromethyl)-1H-pyrazol-3-yl]thiophene-2-carboxylic acid
5-[1-Methyl-5-(trifluoromethyl)-1H-pyrazol-3-yl]thiophene-2-carboxylic acid
Formel: C₁₀H₇F₃N₂O₂S
Molecular Weight: 276.235 g/mol
Schmelzpunkt: 212…215 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00206674
CAS-Nummer: 223499-20-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR