2-Bromo-5-(difluoromethyl)thiophene

Lieferant: Apollo Scientific
Achtung

PC49250-250MG PC49250-500MG
APOSPC49250-250MGEA 0 EUR
APOSPC49250-250MG APOSPC49250-500MG
2-Bromo-5-(difluoromethyl)thiophene
2-Bromo-5-(difluoromethyl)thiophene
Formel: C₅H₃BrF₂S
Lagertemperatur: Kühlschrank
MDL: MFCD22378823
CAS-Nummer: 924646-90-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR