5-Bromo-3-fluoro-N1-methylbenzene-1,2-diamine

Lieferant: Apollo Scientific
Achtung

PC49259-1G PC49259-250MG
APOSPC49259-1GEA 574 EUR
APOSPC49259-1G APOSPC49259-250MG
5-Bromo-3-fluoro-N1-methylbenzene-1,2-diamine
5-Bromo-3-fluoro-N1-methylbenzene-1,2-diamine
Formel: C₇H₈BrFN₂
Lagertemperatur: Kühlschrank
MDL: MFCD22415278
CAS-Nummer: 1396503-78-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR