2-[3-Chlor-5-(trifluormethyl)-2-pyridyl]-2-(2,6-dichlorphenyl)acetonitril 97%

Lieferant: Apollo Scientific

PC4926-1G PC4926-500MG PC4926-5G
APOSPC4926-1GEA 275 EUR
APOSPC4926-1G APOSPC4926-500MG APOSPC4926-5G
2-[3-Chlor-5-(trifluormethyl)-2-pyridyl]-2-(2,6-dichlorphenyl)acetonitril 97%
2-[3-Chlor-5-(trifluormethyl)-2-pyridyl]-2-(2,6-dichlorphenyl)acetonitril
Formel: C₁₄H₆Cl₃F₃N₂
Schmelzpunkt: 107…109 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00172210
CAS-Nummer: 338407-06-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR