4-Bromo-2-(trifluoromethoxy)benzotrifluoride 95%

Lieferant: Apollo Scientific

PC49263-1G PC49263-250MG
APOSPC49263-1GEA 0 EUR
APOSPC49263-1G APOSPC49263-250MG
4-Bromo-2-(trifluoromethoxy)benzotrifluoride 95%
4-Bromo-2-(trifluoromethoxy)benzotrifluoride
Formel: C₈H₃BrF₆O
Lagertemperatur: Raumtemperatur
MDL: MFCD22415279
CAS-Nummer: 1396503-79-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR