Ethyl 3-amino-7-(trifluoromethyl)benzo[b]thiophene-2-carboxylate

Lieferant: Apollo Scientific
Achtung

PC49266-1G
APOSPC49266-1GEA 0 EUR
APOSPC49266-1G
Ethyl 3-amino-7-(trifluoromethyl)benzo[b]thiophene-2-carboxylate
Ethyl 3-amino-7-(trifluoromethyl)benzo[b]thiophene-2-carboxylate
Formel: C₁₂H₁₀F₃NO₂S
Schmelzpunkt: 177…178 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD21602381
CAS-Nummer: 1396503-80-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR