2-Hydroxyphenylsulphur pentafluoride

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-(Pentafluorosulphanyl)phenol, 2-(Pentafluorothio)phenol

PC49268-1G
APOSPC49268-1GEA 364 EUR
APOSPC49268-1G
2-Hydroxyphenylsulphur pentafluoride
2-Hydroxyphenylsulphur pentafluoride
Formel: C₆H₅F₅OS
Molecular Weight: 220.16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22415280
CAS-Nummer: 1126968-75-9
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR