4-Fluor-3-(trifluormethoxy)benzylbromid 98%

Lieferant: Apollo Scientific
Gefahr

PC49274-1G PC49274-5G PC49274-10G
APOSPC49274-1GEA 0 EUR
APOSPC49274-1G APOSPC49274-5G APOSPC49274-10G
4-Fluor-3-(trifluormethoxy)benzylbromid 98%
4-Fluor-3-(trifluormethoxy)benzylbromid
Formel: C₈H₅BrF₄O
Lagertemperatur: Raumtemperatur
MDL: MFCD06660256
CAS-Nummer: 86256-50-0
UN: 3265
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR